Learn More
Aviva Systems Biology Corp TRIM24 Antibody - middle region (ARP38391_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38391_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM24. Alternate names: PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56634