Learn More
Aviva Systems Biology Corp TRIM25 Antibody - middle region (ARP58106_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58106_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM25. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TRIM25. Alternate names: EFP, RNF147, Z147, ZNF147ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-286-437