Learn More
ABclonal Technology TRIM25 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1293820UL
Encompass_Preferred
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein is an RNA binding protein, functions as a ubiquitin E3 ligase and is involved in multiple cellular processes, including regulation of antiviral innate immunity.
Catalog No. 50-000-98267