Learn More
Aviva Systems Biology Corp TRIM26 Antibody - N-terminal region (ARP34178_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34178_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM26. Alternate names: DASS-211F20.5, AFP, RNF95, ZNF173ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55284