Learn More
Aviva Systems Biology Corp TRIM27 Antibody - middle region (ARP34701_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34701_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM27. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM27. Alternate names: RFP, RNF76ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55520