Learn More
Aviva Systems Biology Corp TRIM28 Antibody - C-terminal region (ARP38681_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38681_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM28. Alternate names: KAP1, TF1B, RNF96, TIF1BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56753