Learn More
Aviva Systems Biology Corp TRIM28 Antibody - N-terminal region (ARP31460_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31460_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM28. Alternate names: FLJ29029, KAP1, RNF96, TF1B, TIF1BApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-54233