Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 23650MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: TRIM29/1041. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: A431 cells. Tonsil or Squamous cell carcinoma. Cellular Staining: Cytoplasmic & Cell Surface. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment (126 Amino acid residues between aa 1-200) of human TRIM29 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 66kDa.Chromosome: 11q23.3. Gene Symbol: TRIM29. Gene ID: 23650. Ensemble: ENSG00000137699. Uniprot: XP_016872943.
Catalog No. 50-188-4709