Learn More
Aviva Systems Biology Corp TRIM29 Antibody - middle region (ARP32521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM29. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM29. Alternate names: ATDC, FLJ36085ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54633