Learn More
Aviva Systems Biology Corp TRIM3 Antibody - N-terminal region (ARP34698_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34698_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM3. Alternate names: BERP, HAC1, RNF22, RNF97ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55518