Learn More
Aviva Systems Biology Corp TRIM31 Antibody - middle region (ARP57948_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57948_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM31. Alternate names: C6orf13, HCG1, HCGI, RNFApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-62186