Learn More
Aviva Systems Biology Corp TRIM31 Antibody - N-terminal region (ARP34717_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34717_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM31. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM31. Alternate names: RNF, HCG1, HCGI, C6orf13ApplicationIHC, WB. Species reactivity: Cow, Human, Mouse, Rabbit
Catalog No. 50-014-55530