Learn More
Aviva Systems Biology Corp TRIM32 Antibody - C-terminal region (ARP38966_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38966_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM32. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM32. Alternate names: BBS11, HT2A, LGMD2H, TATIPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56871