Learn More
Aviva Systems Biology Corp TRIM33 Antibody - middle region (ARP39214_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39214_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM33. Alternate names: FLJ32925, PTC7, RFG7, TF1G, TIF1G, TIF1GAMMA, TIFGAMMA, ECTOApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56944