Learn More
Aviva Systems Biology Corp TRIM34 Antibody - N-terminal region (ARP34757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM34. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM34. Alternate names: IFP1, RNF21ApplicationWB. Species reactivity: Horse, Human, Rat
Catalog No. 50-014-55549