Learn More
Aviva Systems Biology Corp TRIM35 Antibody - C-terminal region (ARP34872_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34872_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM35. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM35. Alternate names: HLS5, MAIRApplicationWB. Species reactivity: Human
Catalog No. 50-014-55593