Learn More
Aviva Systems Biology Corp TRIM36 Antibody - middle region (ARP42989_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42989_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM36. Alternate names: HAPRIN, RBCC728, RNF98ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58384