Learn More
Aviva Systems Biology Corp Trim36 Antibody - N-terminal region (ARP43257_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43257_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Trim36. It was validated on Western Blot. . Alternate names: Trim36ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-065