Learn More
Aviva Systems Biology Corp TRIM38 Antibody - N-terminal region (ARP38787_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38787_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM38. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM38. Alternate names: RNF15, RORETApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-53114