Learn More
Aviva Systems Biology Corp TRIM38 Antibody - middle region (ARP57852_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57852_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM38. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TRIM38. Alternate names: MGC8946, RNF15, RORETApplicationWB. Species reactivity: Human
Catalog No. 50-286-404