Learn More
Aviva Systems Biology Corp TRIM39 Antibody - N-terminal region (ARP34755_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34755_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM39. Alternate names: TFP, RNF23, TRIM39BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55547