Learn More
Aviva Systems Biology Corp TRIM39 Antibody - middle region (ARP34756_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34756_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM39. Alternate names: MGC32984, RNF23, TFP, TRIM39BApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55548