Learn More
Aviva Systems Biology Corp TRIM4 Antibody - N-terminal region (ARP34889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM4. Alternate names: RNF87ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55601