Learn More
Aviva Systems Biology Corp TRIM41 Antibody - middle region (ARP34893_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34893_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM41. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM41. Alternate names: MGC1127, MGC31991, RINCKApplicationWB. Species reactivity: Human
Catalog No. 50-014-55603