Learn More
Aviva Systems Biology Corp TRIM41 Antibody - N-terminal region (ARP34763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM41. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM41. Alternate names: MGC1127, MGC31991, RINCKApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55551