Learn More
Aviva Systems Biology Corp TRIM41 Antibody - C-terminal region (ARP34482_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34482_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM41. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM41. Alternate names: MGC1127, MGC31991, RINCKApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55412