Learn More
Aviva Systems Biology Corp TRIM45 Antibody - middle region (ARP43331_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43331_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM45. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM45. Alternate names: FLJ13181, RNF99ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58499