Learn More
Aviva Systems Biology Corp TRIM48 Antibody - C-terminal region (ARP51681_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM48. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM48. Alternate names: MGC4827, RNF101ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-60586