Learn More
Aviva Systems Biology Corp TRIM49 Antibody - middle region (ARP58057_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58057_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM49. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM49. Alternate names: RNF18, TRIM49L2ApplicationWB. Species reactivity: Human
Catalog No. 50-286-432