Learn More
Aviva Systems Biology Corp TRIM5 Antibody - N-terminal region (ARP39741_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39741_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM5. Alternate names: RNF88, TRIM5alphaApplicationWB. Species reactivity: Human
Catalog No. 50-014-57162