Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TRIM5 Human N-His 100ug
SDP

Catalog No. 5000195433
Encompass_Preferred

TRIM5 Protein, Human (N-His) is a recombinant fragment of human TRIM5 (residues M1-Q248) expressed in Escherichia coli and supplied with an N-terminal 6xHis affinity tag. The protein is purified to greater than 95% purity, has an approximate molecular weight of 30 kDa, and is intended for biochemical assays, binding studies, antibody production, and structural analyses. Store frozen and consult the certificate of analysis for batch-specific details.

  • Recombinant human sequence corresponding to residues M1-Q248
  • N-terminal 6xHis tag for affinity purification
  • Expressed in E. coli for high-yield production
  • Purity greater than 95% as determined by reducing SDS-PAGE
  • Approximate molecular weight ~30 kDa
  • Stable when stored frozen and suitable for short-term 4°C handling

Catalog No. 50-001-95433 Supplier Medchemexpress LLC Supplier No. HYP71385A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TRIM5 Protein, Human (N-His) is a recombinant fragment of human TRIM5 (residues M1-Q248) expressed in Escherichia coli and supplied with an N-terminal 6xHis affinity tag. The protein is purified to greater than 95% purity, has an approximate molecular weight of 30 kDa, and is intended for biochemical assays, binding studies, antibody production, and structural analyses. Store frozen and consult the certificate of analysis for batch-specific details.

  • Recombinant human sequence corresponding to residues M1-Q248
  • N-terminal 6xHis tag for affinity purification
  • Expressed in E. coli for high-yield production
  • Purity greater than 95% as determined by reducing SDS-PAGE
  • Approximate molecular weight ~30 kDa
  • Stable when stored frozen and suitable for short-term 4°C handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.