Learn More
Medchemexpress LLC TRIM5 Human N-His 50ug
SDP
Catalog No. 5000195962
Encompass_Preferred
Recombinant human TRIM5 protein (N-terminal 6xHis tag, residues M1-Q248) produced in Escherichia coli and provided lyophilized. The protein functions as a capsid-specific restriction factor that activates E3 ubiquitin ligase activity and innate immune signaling, and is intended for research into viral restriction, ubiquitination, NF-κB/MAPK signaling, and autophagy.
  • Expressed in Escherichia coli with an N-terminal 6xHis tag for affinity purification.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • High purity (>95% by reducing SDS-PAGE) suitable for biochemical and signaling assays.
  • Includes TRIM5 fragment corresponding to residues M1-Q248, approximately 30 kDa.
  • Supports studies of viral capsid recognition, ubiquitination, and autophagy pathways.
Catalog No. 50-001-95962 Supplier Medchemexpress LLC Supplier No. HYP71385A50UG
Recombinant human TRIM5 protein (N-terminal 6xHis tag, residues M1-Q248) produced in Escherichia coli and provided lyophilized. The protein functions as a capsid-specific restriction factor that activates E3 ubiquitin ligase activity and innate immune signaling, and is intended for research into viral restriction, ubiquitination, NF-κB/MAPK signaling, and autophagy.
  • Expressed in Escherichia coli with an N-terminal 6xHis tag for affinity purification.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • High purity (>95% by reducing SDS-PAGE) suitable for biochemical and signaling assays.
  • Includes TRIM5 fragment corresponding to residues M1-Q248, approximately 30 kDa.
  • Supports studies of viral capsid recognition, ubiquitination, and autophagy pathways.