Learn More
Medchemexpress LLC TRIM5 Human N-His 5ug
SDP
Catalog No. 5000195650
Encompass_Preferred
Recombinant human TRIM5 protein (residues M1-Q248) supplied as a lyophilized powder with an N-terminal 6xHis tag, expressed in E. coli. Designed for biochemical and cell biology applications, the material is provided at 5 μg with >95% purity and an approximate molecular weight of 30 kDa; follow the datasheet for reconstitution and storage instructions.
Catalog No. 50-001-95650 Supplier Medchemexpress LLC Supplier No. HYP71385A5UG
Recombinant human TRIM5 protein (residues M1-Q248) supplied as a lyophilized powder with an N-terminal 6xHis tag, expressed in E. coli. Designed for biochemical and cell biology applications, the material is provided at 5 μg with >95% purity and an approximate molecular weight of 30 kDa; follow the datasheet for reconstitution and storage instructions.