Learn More
Aviva Systems Biology Corp TRIM55 Antibody - N-terminal region (ARP43373_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43373_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM55. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM55. Alternate names: MURF-2, RNF29, muRF2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58511