Learn More
Aviva Systems Biology Corp TRIM58 Antibody - middle region (ARP57993_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57993_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM58. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM58. Alternate names: BIA2, DKFZp434C091ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62205