Learn More
Aviva Systems Biology Corp TRIM60 Antibody - N-terminal region (ARP43434_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM60. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM60. Alternate names: FLJ35882, MGC119325, RNF129, RNF33ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58532