Learn More
Aviva Systems Biology Corp TRIM62 Antibody - N-terminal region (ARP34576_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34576_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM62. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM62. Alternate names: FLJ10759, FLJ16558, DEAR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55459