Learn More
ABclonal Technology TRIM62 Rabbit pAb
SDP
Catalog No. 5000105712
Encompass_Preferred
Enables identical protein binding activity, transcription coactivator activity, and ubiquitin-protein transferase activity. Involved in several processes, including negative regulation of viral transcription, positive regulation of NF-kappaB transcription factor activity, and positive regulation of antifungal innate immune response. Is active in cytoplasm.
Catalog No. 50-001-05712 Supplier ABclonal Technology Supplier No. A158551000UL
Enables identical protein binding activity, transcription coactivator activity, and ubiquitin-protein transferase activity. Involved in several processes, including negative regulation of viral transcription, positive regulation of NF-kappaB transcription factor activity, and positive regulation of antifungal innate immune response. Is active in cytoplasm.