Learn More
Aviva Systems Biology Corp TRIM63 Antibody - middle region (ARP43365_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43365_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM63. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM63. Alternate names: FLJ32380, IRF, MURF1, MURF2, RNF28, SMRZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53856