Learn More
Aviva Systems Biology Corp TRIM67 Antibody - C-terminal region (ARP42961_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42961_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM67. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM67. Alternate names: FLJ44831, TNLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58374