Learn More
Aviva Systems Biology Corp TRIM68 Antibody - middle region (ARP39328_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39328_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM68. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM68. Alternate names: FLJ10369, MGC126176, RNF137, SS-56, SS56, GC109ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56987