Learn More
Aviva Systems Biology Corp TRIM69 Antibody - N-terminal region (ARP34764_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34764_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM69. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM69. Alternate names: HSD34, RNF36, TrifApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55552