Learn More
Aviva Systems Biology Corp TRIM8 Antibody - N-terminal region (ARP39655_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39655_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM8. Alternate names: GERP, RNF27ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57123