Learn More
Aviva Systems Biology Corp TRIM9 Antibody - C-terminal region (ARP34861_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34861_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM9. Alternate names: RNF91, SPRINGApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-55588