Learn More
MSC TRIMACO Shoe Guards, Blue, Non-Skid, Poly, Universal, 10 Pair
Catalog No. 5000767485
Encompass
These shoe guards are designed to protect floors and keep footwear clean. They feature a non-skid sole for enhanced safety.
  • Made from polyethylene material
  • Universal size for most footwear
  • Non-skid sole for improved traction
  • Comes in a pack of 10 pairs
  • Individually bagged for hygiene
Catalog No. 50-007-67485 Supplier MSC Supplier No. 61738076
These shoe guards are designed to protect floors and keep footwear clean. They feature a non-skid sole for enhanced safety.
  • Made from polyethylene material
  • Universal size for most footwear
  • Non-skid sole for improved traction
  • Comes in a pack of 10 pairs
  • Individually bagged for hygiene