Learn More
Medchemexpress LLC Trimagnesium dicitra 1kg
SDP
Catalog No. 5000206062
Encompass_Preferred
Trimagnesium dicitrate nonahydrate 99% (Magnesium citrate tribasic nonahydrate 99%) is an orally active magnesium salt of citric acid in the nonahydrate form Trimagnesium dicitrate nonahydrate 99% used as a stabilizer and anticaking agent in food supplements in solid and chewable forms[1]
Catalog No. 50-002-06062 Supplier Medchemexpress LLC Supplier No. HYW1157981KG
Trimagnesium dicitrate nonahydrate 99% (Magnesium citrate tribasic nonahydrate 99%) is an orally active magnesium salt of citric acid in the nonahydrate form Trimagnesium dicitrate nonahydrate 99% used as a stabilizer and anticaking agent in food supplements in solid and chewable forms[1]