Learn More
Medchemexpress LLC Trimagnesium dicitra 500g
SDP
Catalog No. 5000206493
Encompass_Preferred
Trimagnesium dicitrate nonahydrate 99% (Magnesium citrate tribasic nonahydrate 99%) is an orally active magnesium salt of citric acid in the nonahydrate form Trimagnesium dicitrate nonahydrate 99% used as a stabilizer and anticaking agent in food supplements in solid and chewable forms[1]
Catalog No. 50-002-06493 Supplier Medchemexpress LLC Supplier No. HYW115798500G
Trimagnesium dicitrate nonahydrate 99% (Magnesium citrate tribasic nonahydrate 99%) is an orally active magnesium salt of citric acid in the nonahydrate form Trimagnesium dicitrate nonahydrate 99% used as a stabilizer and anticaking agent in food supplements in solid and chewable forms[1]