Learn More
Electron Microscopy Sciences Trimaide Razor Blade Holder
SDP
Catalog No. 50949405
Encompass_Preferred
A razor blade holder for trimming blocks. Holds all types of razors. It gives you a better hold on the blade while you trim your blocks. The blade slides sideways in the holder allowing use of the entire edge. IT PROTECTS FINGERS!!!
Catalog No. 50-949-405 Supplier Electron Microscopy Sciences Supplier No. 72560
A razor blade holder for trimming blocks. Holds all types of razors. It gives you a better hold on the blade while you trim your blocks. The blade slides sideways in the holder allowing use of the entire edge. IT PROTECTS FINGERS!!!