Learn More
TARGETMOL CHEMICALS INC Trimebutine CTB salt 1MG
SDP
Catalog No. 502879455
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trimebutine CTB salt (GIC-1001) is an opioid receptor agonist used in the study of pain. Purity 99.53%
Catalog No. 50-287-9455 Supplier TARGETMOL CHEMICALS INC Supplier No. T274131MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Trimebutine CTB salt (GIC-1001) is an opioid receptor agonist used in the study of pain. Purity 99.53%