Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimellitic Anhydride Acid Chloride 99%, Thermo Scientific™

Catalog No. AC139881000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC139881000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139881000 Supplier Thermo Scientific Chemicals Supplier No. 139881000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1204-28-0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C9H3ClO4
Quantity 100g
MDL Number MFCD00005924
Synonym trimellitic anhydride chloride, 4-chloroformylphthalic anhydride, 4-chloroformyl phthalic anhydride, trimellitic acid anhydride chloride, anhydrotrimellitic acid chloride, 5-isobenzofurancarbonyl chloride, 1,3-dihydro-1,3-dioxo, trimellitic anhydride acid chloride, 4-chlorocarbonyl phthalic anhydride, 1,3-benzofurandione-5-carbonyl chloride, trimellitic anhydride monochloride
InChI Key NJMOHBDCGXJLNJ-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1C(=O)Cl)C(=O)OC2=O
IUPAC Name 1,3-dioxo-2-benzofuran-5-carbonyl chloride
Molecular Weight (g/mol) 210.569
PubChem CID 70998
Formula Weight 210.57
Percent Purity ≥98.5%
Chemical Name or Material Trimellitic anhydride acid chloride, 99%
Show More Show Less
EINECSNumber 214-874-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.